cis-β-Methylstyrene ≥98% stabilised

Supplier: Ambeed
Danger

Synonyms: cis-1-Phenyl-1-propene, cis-Propenylbenzene

Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

A204458-1G A204458-5G A204458-10G
77550-314EA 59 USD
77550-314 77550-316 77879-277
cis-β-Methylstyrene ≥98% stabilised
cis-beta-Methylstyrene

Precaución: for Research Use Only

Formula: C₉H₁₀
MDL Number: MFCD00065137
CAS Number: 766-90-5
UN: 3295
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR