Citric acid ≥98%

Supplier: Ambeed
Warning

A198670-100G A198670-10G A198670-25G A198670-500G
77550-262EA 8 USD
77550-262 77593-282 77593-284 77593-286
Citric acid ≥98%
Citric acid

Precaución: for Research Use Only

Formula: HOC(COOH)(CH₂COOH)₂
MDL Number: MFCD00011669
CAS Number: 77-92-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR