1,3,5-Trifluorobenzene ≥98%

Supplier: Ambeed
Danger

A196792-25G A196792-5G
77550-252EA 55.62 USD
77550-252 77615-666
1,3,5-Trifluorobenzene ≥98%
1,3,5-Trifluorobenzene

Precaución: for Research Use Only

Formula: C₆H₃F₃
MW: 132.09 g/mol
MDL Number: MFCD00000333
CAS Number: 372-38-3
UN: 1993
ADR: 3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR