t-Boc-N-Amido-PEG2-acid 95%

Supplier: Ambeed
Warning

Synonyms: 2,2-Dimethyl-4-oxo-3,8,11-trioxa-5-azatetradecan-14-oic acid, 3-[2-(2-([(tert-Butoxy)carbonyl]amino)ethoxy)ethoxy]propanoic acid, Boc-NH-PEG2-CH2CH2COOH, N-Boc-3-[2-(2-aminoethoxy)ethoxy]propionic Acid

A192040-1G A192040-5G A192040-250MG A192040-25G A192040-100MG
77550-230EA 17 USD
77550-230 77550-232 77567-788 77611-838 77892-237
t-Boc-N-Amido-PEG2-acid 95%
t-Boc-N-Amido-PEG2-acid

Precaución: For research use only.

Formula: C₁₂H₂₃NO₆
MDL Number: MFCD22056304
CAS Number: 1365655-91-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR