LCL161 ≥98%

Supplier: Ambeed
Warning

Synonyms: (2S)-N-[(1S)-1-cyclohexyl-2-[(2S)-2-[4-(4-fluorobenzoyl)-1,3-thiazol-2-yl]pyrrolidin-1-yl]-2-oxoethyl]-2-(methylamino)propenamide, XIAP Inhibitor, lcl161, LCL-161, IAP inhibitor, (S)-N-((S)-1-Cyclohexyl-2-((S)-2-(4-(4-fluorobenzoyl)thiazol-2-yl)pyrrolidin-1-yl)-2-oxoethyl)-2-(methylamino)propanamide

A188719-100MG A188719-25MG A188719-50MG A188719-1MG
77550-204EA 879.62 USD
77550-204 77550-206 77550-208 77615-598
LCL161 ≥98%
LCL161

Precaución: for Research Use Only

Formula: C₂₆H₃₃FN₄O₃S
MW: 500.63 g/mol
MDL Number: MFCD23160049
CAS Number: 1005342-46-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR