Methyl 6-chloro-3-fluoropicolinate ≥97%

Supplier: Ambeed
Warning

A186590-1G A186590-250MG A186590-100MG A186590-5G
77550-194EA 204 USD
77550-194 77550-196 77671-638 77883-391
Methyl 6-chloro-3-fluoropicolinate ≥97%
Methyl 6-chloro-3-fluoropicolinate

Precaución: for Research Use Only

Formula: C₇H₅ClFNO₂
MW: 189.57 g/mol
MDL Number: MFCD14698146
CAS Number: 1030829-21-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR