1H-Pyrazol-1-ol 97%

Supplier: Ambeed
Warning

A180234-1G A180234-10G
77550-164EA 25.75 USD
77550-164 77567-654
1H-Pyrazol-1-ol 97%
1H-Pyrazol-1-ol

Precaución: For research use only.

Formula: C₃H₄N₂O
MW: 84.08 g/mol
MDL Number: MFCD11850516
CAS Number: 81945-73-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR