2,4-Dichloro-5-(trifluoromethyl)pyrimidine 98%

Supplier: Ambeed
Danger

A178973-25G A178973-10G
77550-160EA 48.41 USD
77550-160 77567-640
2,4-Dichloro-5-(trifluoromethyl)pyrimidine 98%
2,4-Dichloro-5-(trifluoromethyl)pyrimidine

Precaución: For research use only.

Formula: C₅HCl₂F₃N₂
MW: 216.98 g/mol
MDL Number: MFCD03426408
CAS Number: 3932-97-6
UN: 2922
ADR: 8,6.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR