2,6-Dichloropyridine-4-sulfonyl chloride ≥95%

Supplier: Ambeed
Danger

A177595-1G A177595-100MG A177595-5G
77550-140EA 400 USD
77550-140 77748-161 77748-162
2,6-Dichloropyridine-4-sulfonyl chloride ≥95%
2,6-Dichloropyridine-4-sulfonyl chloride

Precaución: for Research Use Only

Formula: C₅H₂Cl₃NO₂S
MDL Number: MFCD18411528
CAS Number: 1058741-91-5
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR