Sulfamoyl chloride ≥98%

Supplier: Ambeed
Danger

A173702-1G A173702-25G A173702-5G
77550-122EA 5 USD
77550-122 77550-124 77550-126
Sulfamoyl chloride ≥98%
Sulfamoyl chloride

Precaución: for Research Use Only

Formula: ClH₂NO₂S
MDL Number: MFCD11519990
CAS Number: 7778-42-9
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR