(Phenylthio)trimethylsilane ≥95%

Supplier: Ambeed
Danger

Synonyms: Phenylthiotrimethylsilane

A173471-25G A173471-5G
77550-116EA 242.05 USD
77550-116 77550-118
(Phenylthio)trimethylsilane ≥95%
(Phenylthio)trimethylsilane

Precaución: for Research Use Only

Formula: C₉H₁₄SSi
MW: 182.36 g/mol
MDL Number: MFCD00008272
CAS Number: 4551-15-9
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR