5-Nitro-2-furaldehyde 98%

Supplier: Ambeed
Danger

Synonyms: 5-Nitrofurfural

A169721-25G A169721-10G
77550-092EA 37.08 USD
77550-092 77567-492
5-Nitro-2-furaldehyde 98%
5-Nitro-2-furaldehyde

Precaución: For research use only.

Formula: C₅H₃NO₄
MW: 141.08 g/mol
MDL Number: MFCD00003230
CAS Number: 698-63-5
UN: 1325
ADR: 4.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR