(SP-4-2)-Bromo[dicyclohexyl[3-(1,1-dimethylethoxy)-6-methoxy-2',6'-bis(1-methylethyl)[1,1'-biphenyl]-2-yl-κC1']phosphine-κP][4-[[2-(trimethylsilyl)ethoxy]carbonyl]phenyl]Palladium 95%

Supplier: Ambeed
Warning

A1683265-1G A1683265-250MG A1683265-100MG A1683265-5G
77550-080EA 677 USD
77550-080 77550-082 77567-462 77567-464
(SP-4-2)-Bromo[dicyclohexyl[3-(1,1-dimethylethoxy)-6-methoxy-2',6'-bis(1-methylethyl)[1,1'-biphenyl]-2-yl-κC1']phosphine-κP][4-[[2-(trimethylsilyl)ethoxy]carbonyl]phenyl]Palladium 95%
(SP-4-2)-Bromo[dicyclohexyl[3-(1,1-dimethylethoxy)-6-methoxy-2',6'-bis(1-methylethyl)[1,1'-biphenyl]-2-yl-κC1']phosphine-κP][4-[[2-(trimethylsilyl)ethoxy]carbonyl]phenyl]Palladium

Precaución: For research use only.

MW: 944.440 g/mol
CAS Number: 2489525-81-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR