3,4-Diethylhexane-3,4-diol 95%

Supplier: Ambeed
Warning

A1635726-25G A1635726-5G A1635726-1G A1635726-100G
77550-034EA 173 USD
77550-034 77550-036 77567-388 77876-483
3,4-Diethylhexane-3,4-diol 95%
3,4-Diethylhexane-3,4-diol

Precaución: For research use only.

Formula: C₁₀H₂₂O₂
MDL Number: MFCD01723418
CAS Number: 6931-71-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR