Ethyl 8-bromo-5-methyl-6-oxo-5,6-dihydro-4H-benzo[f]imidazo[1,5-a][1,4]diazepine-3-carboxylate 98%

Supplier: Ambeed
Warning

A162173-250MG A162173-1G
77550-030EA 281.19 USD
77550-030 77567-370
Ethyl 8-bromo-5-methyl-6-oxo-5,6-dihydro-4H-benzo[f]imidazo[1,5-a][1,4]diazepine-3-carboxylate 98%
Ethyl 8-bromo-5-methyl-6-oxo-5,6-dihydro-4H-benzo[f]imidazo[1,5-a][1,4]diazepine-3-carboxylate

Precaución: For research use only.

Formula: C₁₅H₁₄BrN₃O₃
MW: 364.19 g/mol
MDL Number: MFCD12911997
CAS Number: 78756-37-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR