5-Hydroxyindole 97%

Supplier: Ambeed
Warning

Synonyms: 1H-Indol-5-ol, 5-Indolol, Indol-5-ol, 5-Hydroxy-1H-indole

A161216-10G A161216-5G A161216-1G A161216-25G A161216-100G
77550-020EA 64.89 USD
77550-020 77567-350 77615-314 77615-316 77746-100
5-Hydroxyindole 97%
5-Hydroxyindole

Precaución: For research use only.

Formula: C₈H₇NO
MW: 133.15 g/mol
MDL Number: MFCD00005677
CAS Number: 1953-54-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR