5-Phenyloxazolidin-2-one ≥97%

Supplier: Ambeed
Warning

A160323-250MG A160323-5G
77550-014EA 18 USD
77550-014 77871-803
5-Phenyloxazolidin-2-one ≥97%
5-Phenyloxazolidin-2-one

Precaución: for Research Use Only

Formula: C₉H₉NO₂
MDL Number: MFCD01747962
CAS Number: 7693-77-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR