(S)-(+)-Camptothecin ≥98%

Supplier: Ambeed
Danger

Synonyms: (S)-4-Ethyl-4-hydroxy-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione, (+)-Camptothecin, 4-Ethyl-4-hydroxy-1H-pyrano-[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione

A159794-1G A159794-5G A159794-100G A159794-10G A159794-25G A159794-1MG
77550-010EA 15.45 USD
77550-010 77550-012 77611-548 77611-550 77611-552 77685-307
(S)-(+)-Camptothecin ≥98%
(S)-(+)-Camptothecin

Precaución: for Research Use Only

Formula: C₂₀H₁₆N₂O₄
MW: 348.36 g/mol
MDL Number: MFCD00081076
CAS Number: 7689-03-4
UN: 1544
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR