Coumarin boronic acid 95+%

Supplier: Ambeed
Warning

Synonyms: (2-Oxochromen-7-yl)boronic acid, CBA, B-(2-oxo-2H-1-benzopyran-7-yl)-boronic acid

A155344-100MG A155344-250MG
77549-972EA 114 USD
77549-972 77685-282
Coumarin boronic acid 95+%
Coumarin boronic acid

Precaución: for Research Use Only

Formula: C₉H₇BO₄
MW: 189.96 g/mol
MDL Number: MFCD25562900
CAS Number: 1357078-03-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR