Methanesulfonato[Dicyclohexyl[3-(1,1-dimethylethoxy)-6-methoxy-2',6'-bis(1-methylethyl)[1,1'-biphenyl]-2-yl]phosphine](2'-amino-1,1'-biphenyl-2-yl)palladium(II) ≥95%

Supplier: Ambeed
Warning

A1547860-250MG A1547860-1G A1547860-100MG A1547860-50MG A1547860-5G
77549-962EA 418 USD
77549-962 77625-380 77876-262 77876-263 77876-264
Methanesulfonato[Dicyclohexyl[3-(1,1-dimethylethoxy)-6-methoxy-2',6'-bis(1-methylethyl)[1,1'-biphenyl]-2-yl]phosphine](2'-amino-1,1'-biphenyl-2-yl)palladium(II) ≥95%
Methanesulfonato[Dicyclohexyl[3-(1,1-dimethylethoxy)-6-methoxy-2',6'-bis(1-methylethyl)[1,1'-biphenyl]-2-yl]phosphine](2'-amino-1,1'-biphenyl-2-yl)palladium(II)

Precaución: for Research Use Only

Formula: C₄₈H₆₆NO₅PPdS
MW: 906.5 g/mol
CAS Number: 2489525-82-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR