4-Oxo-4H-pyrido[1,2-a]pyrimidine-2-carboxylic acid hydrochloride 98%

Supplier: Ambeed
Warning

A1544101-5G A1544101-25G
77549-956EA 186.43 USD
77549-956 77567-250
4-Oxo-4H-pyrido[1,2-a]pyrimidine-2-carboxylic acid hydrochloride 98%
4-Oxo-4H-pyrido[1,2-a]pyrimidine-2-carboxylic acid hydrochloride

Precaución: For research use only.

Formula: C₉H₇ClN₂O₃
MW: 226.61 g/mol
CAS Number: 2499661-06-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR