(1S,2R)-2-(Methoxycarbonyl)cyclopropanecarboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: (1S,2R)-rel-2-(methoxycarbonyl)cyclopropane-1-carboxylic acid

A153858-5G A153858-1G A153858-100MG A153858-250MG A153858-25G
77549-952EA 80.34 USD
77549-952 77567-236 77615-256 77748-058 77748-059
(1S,2R)-2-(Methoxycarbonyl)cyclopropanecarboxylic acid 98%
(1S,2R)-2-(Methoxycarbonyl)cyclopropanecarboxylic acid

Precaución: For research use only.

Formula: C₆H₈O₄
MW: 144.12 g/mol
MDL Number: MFCD21332871
CAS Number: 31420-47-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR