Methyl 3-bromocyclobutane-1-carboxylate ≥98%

Supplier: Ambeed
Warning

A153529-1G A153529-5G
77549-944EA 48 USD
77549-944 77892-047
Methyl 3-bromocyclobutane-1-carboxylate ≥98%
Methyl 3-bromocyclobutane-1-carboxylate

Precaución: for Research Use Only

Formula: C₆H₉BrO₂
MDL Number: MFCD20625735
CAS Number: 4935-00-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR