3-(5-Fluoro-1-oxoisoindolin-2-yl)piperidine-2,6-dione 98%

Supplier: Ambeed
Warning

A1534164-1G A1534164-250MG
77549-936EA 251.32 USD
77549-936 77567-222
3-(5-Fluoro-1-oxoisoindolin-2-yl)piperidine-2,6-dione 98%
3-(5-Fluoro-1-oxoisoindolin-2-yl)piperidine-2,6-dione

Precaución: For research use only.

Formula: C₁₃H₁₁FN₂O₃
MW: 262.236 g/mol
MDL Number: MFCD34186457
CAS Number: 2468780-76-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR