3-(tert-Butyl)-6-(ethylthio)-1,3,5-triazine-2,4(1H,3H)-dione 97%

Supplier: Ambeed
Warning

A1533355-25G A1533355-5G A1533355-1G
77549-932EA 120.51 USD
77549-932 77567-220 77597-972
3-(tert-Butyl)-6-(ethylthio)-1,3,5-triazine-2,4(1H,3H)-dione 97%
3-(tert-Butyl)-6-(ethylthio)-1,3,5-triazine-2,4(1H,3H)-dione

Precaución: For research use only.

Formula: C₉H₁₅N₃O₂S
MW: 229.29 g/mol
CAS Number: 1360105-53-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR