6-Chloro-2-methyl-2H-indazol-5-amine ≥97%

Supplier: Ambeed
Warning

A1533017-25G A1533017-5G A1533017-1G
77549-928EA 128.75 USD
77549-928 77549-930 77592-874
6-Chloro-2-methyl-2H-indazol-5-amine ≥97%
6-Chloro-2-methyl-2H-indazol-5-amine

Precaución: for Research Use Only

Formula: C₈H₈ClN₃
MW: 181.62 g/mol
CAS Number: 1893125-36-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR