(6-Chloronaphthalen-2-yl)boronic acid 98%

Supplier: Ambeed
Warning

A1527631-250MG A1527631-1G A1527631-100MG
77549-924EA 263 USD
77549-924 77567-214 77876-102
(6-Chloronaphthalen-2-yl)boronic acid 98%
(6-Chloronaphthalen-2-yl)boronic acid
Formula: C₁₀H₈BClO₂
MDL Number: MFCD18411299
CAS Number: 870822-86-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR