Methyl (S)-3-(4-bromothiazol-2-yl)-2-((tert-butoxycarbonyl)amino)propanoate ≥98%

Supplier: Ambeed
Warning

A1527280-1G A1527280-250MG
77549-922EA 318 USD
77549-922 77876-098
Methyl (S)-3-(4-bromothiazol-2-yl)-2-((tert-butoxycarbonyl)amino)propanoate ≥98%
Methyl (S)-3-(4-bromothiazol-2-yl)-2-((tert-butoxycarbonyl)amino)propanoate

Precaución: for Research Use Only

Formula: C₁₂H₁₇BrN₂O₄S
MDL Number: MFCD34179557
CAS Number: 2641451-73-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR