Phenyl((trimethylsilyl)ethynyl)iodonium trifluoromethanesulfonate ≥97%

Supplier: Ambeed
Danger

A1521791-1G A1521791-250MG
77549-914EA 167 USD
77549-914 77876-071
Phenyl((trimethylsilyl)ethynyl)iodonium trifluoromethanesulfonate ≥97%
Phenyl((trimethylsilyl)ethynyl)iodonium trifluoromethanesulfonate

Precaución: for Research Use Only

Formula: C₁₂H₁₄F₃IO₃SSi
CAS Number: 133816-00-9
UN: 2923
ADR: 8,6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR