(2R,3R,5S)-2-(4-Benzamido-2-oxopyrimidin-1(2H)-yl)-5-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)tetrahydrofuran-3-yl (2-cyanoethyl) diisopropylphosphoramidite ≥95%

Supplier: Ambeed
Warning

Synonyms: 3’-dC(Bz)-2’-phosphor amidite

A1512947-1G A1512947-250MG A1512947-100MG
77549-904EA 750.87 USD
77549-904 77597-856 77624-826
(2R,3R,5S)-2-(4-Benzamido-2-oxopyrimidin-1(2H)-yl)-5-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)tetrahydrofuran-3-yl (2-cyanoethyl) diisopropylphosphoramidite ≥95%
(2R,3R,5S)-2-(4-Benzamido-2-oxopyrimidin-1(2H)-yl)-5-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)tetrahydrofuran-3-yl (2-cyanoethyl) diisopropylphosphoramidite

Precaución: for Research Use Only

Formula: C₄₆H₅₂N₅O₈P
MW: 833.907 g/mol
MDL Number: MFCD08062013
CAS Number: 157327-96-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR