tert-Butyl-4-piperazinotetrahydro-1(2H)-pyridinecarboxylate 98%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 4-(piperazin-1-yl)piperidine-1-carboxylate, 4-(Piperazin-1-yl)piperidine, N-BOC protected

A150338-1G A150338-250MG A150338-10G A150338-5G
77549-882EA 91.67 USD
77549-882 77567-152 77600-126 77600-128
tert-Butyl-4-piperazinotetrahydro-1(2H)-pyridinecarboxylate 98%
tert-Butyl-4-piperazinotetrahydro-1(2H)-pyridinecarboxylate

Precaución: For research use only.

Formula: C₁₄H₂₇N₃O₂
MW: 269.39 g/mol
MDL Number: MFCD04973999
CAS Number: 177276-41-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR