((2-Fluoro-8-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalen-1-yl)ethynyl)triisopropylsilane ≥98%

Supplier: Ambeed
Warning

A1502845-1G A1502845-250MG A1502845-100MG A1502845-5G
77549-880EA 66 USD
77549-880 77624-720 77875-820 77875-821
((2-Fluoro-8-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalen-1-yl)ethynyl)triisopropylsilane ≥98%
((2-Fluoro-8-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalen-1-yl)ethynyl)triisopropylsilane

Precaución: for Research Use Only

Formula: C₂₇H₃₈BFO₂Si
MDL Number: MFCD34603716
CAS Number: 2503307-87-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR