(4S,4'S)-1,1'-Bis(3-(tert-butyl)phenyl)-4,4'-diisopropyl-4,4',5,5'-tetrahydro-1H,1'H-2,2'-biimidazole ≥97%

Supplier: Ambeed
Warning

A1501173-100MG A1501173-250MG A1501173-1G
77549-870EA 142 USD
77549-870 77597-764 77624-668
(4S,4'S)-1,1'-Bis(3-(tert-butyl)phenyl)-4,4'-diisopropyl-4,4',5,5'-tetrahydro-1H,1'H-2,2'-biimidazole ≥97%
(4S,4'S)-1,1'-Bis(3-(tert-butyl)phenyl)-4,4'-diisopropyl-4,4',5,5'-tetrahydro-1H,1'H-2,2'-biimidazole

Precaución: for Research Use Only

Formula: C₃₂H₄₆N₄
MDL Number: MFCD34598896
CAS Number: 2374958-80-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR