(S)-2-Amino-N-((S)-1-((4-(hydroxymethyl)phenyl)amino)-1-oxopropan-2-yl)-3-methylbutanamide 97%

Supplier: Ambeed
Warning

Synonyms: Val-Ala-PAB

A1492902-1G A1492902-5G A1492902-250MG A1492902-100MG A1492902-10G
77549-854EA 302.82 USD
77549-854 77567-114 77592-818 77624-520 77624-522
(S)-2-Amino-N-((S)-1-((4-(hydroxymethyl)phenyl)amino)-1-oxopropan-2-yl)-3-methylbutanamide 97%
(S)-2-Amino-N-((S)-1-((4-(hydroxymethyl)phenyl)amino)-1-oxopropan-2-yl)-3-methylbutanamide

Precaución: For research use only.

Formula: C₁₅H₂₃N₃O₃
MW: 293.3614 g/mol
MDL Number: MFCD32641649
CAS Number: 1343476-44-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR