Coenzyme A 85%

Supplier: Ambeed
Warning

Synonyms: (((2R,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-4-hydroxy-2-(((hydroxy((hydroxy((R)-3-hydroxy-2,2-dimethyl-3-((2-((2-sulphanylethyl)carbamoyl)ethyl)carbamoyl)propoxy)phosphoryl)oxy)phosphoryl)oxy)methyl)oxolan-3-yl)oxy)phosphonic acid, CoA

A148168-250MG A148168-100MG
103792-204EA 21 USD
103792-204 103792-206 103799-388 103803-886 103842-000 77549-824 77567-058
Coenzyme A 85%
Coenzyme A

Precaución: For research use only.

Formula: C₂₁H₃₆N₇O₁₆P₃S
MW: 767.53 g/mol
MDL Number: MFCD24368566
CAS Number: 85-61-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR