(S)-11-Benzyl-1-(9H-fluoren-9-yl)-3,6,9,12,15-pentaoxo-2,18-dioxa-4,7,10,13,16-pentaazaicosan-20-oic acid ≥95%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Gly-Gly-Phe-Gly-NH-CH2-O-CH2COOH

A1467104-1G A1467104-250MG A1467104-100MG A1467104-50MG
77549-794EA 897 USD
77549-794 77549-796 77622-206 77875-043
(S)-11-Benzyl-1-(9H-fluoren-9-yl)-3,6,9,12,15-pentaoxo-2,18-dioxa-4,7,10,13,16-pentaazaicosan-20-oic acid ≥95%
(S)-11-Benzyl-1-(9H-fluoren-9-yl)-3,6,9,12,15-pentaoxo-2,18-dioxa-4,7,10,13,16-pentaazaicosan-20-oic acid

Precaución: for Research Use Only

Formula: C₃₃H₃₅N₅O₉
CAS Number: 2264011-98-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR