2,4-Dichloro-5-iodoaniline ≥97%
Supplier: Ambeed
|
Warning
|
A145958-1G
A145958-25G
A145958-5G
77549-776EA
104.03
USD
77549-776
77549-778
77549-780
2,4-Dichloro-5-iodoaniline ≥97%
2,4-Dichloro-5-iodoaniline
Precaución: for Research Use Only
Formula:
C₆H₄Cl₂IN MW: 287.91 g/mol |
MDL Number:
MFCD28786669 CAS Number: 647025-63-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...