2-Indanol ≥98%

Supplier: Ambeed
Warning

Synonyms: 2-Hydroxyindane

A145372-5G A145372-25G
77549-766EA 17.51 USD
77549-766 77748-013
2-Indanol ≥98%
2-Indanol

Precaución: for Research Use Only

Formula: C₉H₁₀O
MW: 134.18 g/mol
MDL Number: MFCD00003800
CAS Number: 4254-29-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR