(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(3-(tert-butoxycarbonyl)phenyl)propanoic acid ≥98%

Supplier: Ambeed
Warning

A1443614-1G A1443614-5G A1443614-100MG
77549-746EA 318 USD
77549-746 77549-748 77874-859
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(3-(tert-butoxycarbonyl)phenyl)propanoic acid ≥98%
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(3-(tert-butoxycarbonyl)phenyl)propanoic acid

Precaución: for Research Use Only

Formula: C₂₉H₂₉NO₆
CAS Number: 210282-33-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR