(S)-10-Benzyl-23-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)-6,9,12,15,18-pentaoxo-3-oxa-5,8,11,14,17-pentaazatricosan-1-oic acid ≥95%

Supplier: Ambeed
Warning

Synonyms: MC-Gly-Gly-Phe-Gly-NH-CH2-O-CH2COOH

A1441480-1G A1441480-250MG A1441480-100MG A1441480-50MG
77549-740EA 550.02 USD
77549-740 77600-078 77597-188 77597-190
(S)-10-Benzyl-23-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)-6,9,12,15,18-pentaoxo-3-oxa-5,8,11,14,17-pentaazatricosan-1-oic acid ≥95%
(S)-10-Benzyl-23-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)-6,9,12,15,18-pentaoxo-3-oxa-5,8,11,14,17-pentaazatricosan-1-oic acid

Precaución: for Research Use Only

Formula: C₂₈H₃₆N₆O₁₀
MW: 616.619 g/mol
CAS Number: 1599440-25-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR