tert-Butyl 4-(piperidin-4-ylmethyl)piperazine-1-carboxylate 98%

Supplier: Ambeed
Danger

Synonyms: tert-butyl 4-(4-piperidylmethyl)piperazine-1-carboxylate

A1441272-1G A1441272-5G A1441272-250MG
77549-736EA 28 USD
77549-736 77549-738 77566-898
tert-Butyl 4-(piperidin-4-ylmethyl)piperazine-1-carboxylate 98%
tert-Butyl 4-(piperidin-4-ylmethyl)piperazine-1-carboxylate

Precaución: For research use only.

Formula: C₁₅H₂₉N₃O₂
MDL Number: MFCD21145604
CAS Number: 1211568-27-2
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR