tert-Butyl 7-oxo-2-azaspiro[4.5]decane-2-carboxylate ≥98%

Supplier: Ambeed
Warning

A143426-250MG A143426-100MG A143426-50MG
77549-732EA 455 USD
77549-732 77891-993 77891-994
tert-Butyl 7-oxo-2-azaspiro[4.5]decane-2-carboxylate ≥98%
tert-Butyl 7-oxo-2-azaspiro[4.5]decane-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₂₃NO₃
MDL Number: MFCD26959114
CAS Number: 1421313-98-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR