2-Bromooxazole 98%

Supplier: Ambeed
Warning

A142825-10G A142825-1G A142825-250MG A142825-5G
77549-726EA 719.97 USD
77549-726 77549-728 77549-730 77566-864
2-Bromooxazole 98%
2-Bromooxazole

Precaución: For research use only.

Formula: C₃H₂BrNO
MW: 147.96 g/mol
MDL Number: MFCD09835085
CAS Number: 125533-82-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR