Fmoc-L-4-carbamoylphe ≥98%

Supplier: Ambeed
Warning

Synonyms: (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-carbamoylphenyl)propanoic acid

A142385-5G A142385-1G A142385-100MG A142385-250MG
77549-722EA 589 USD
77549-722 77592-690 77667-062 77667-064
Fmoc-L-4-carbamoylphe ≥98%
Fmoc-L-4-carbamoylphe

Precaución: for Research Use Only

Formula: C₂₅H₂₂N₂O₅
MDL Number: MFCD01317021
CAS Number: 204716-17-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR