3-Bromo-6-fluoro-2,4-dimethylpyridine ≥97%

Supplier: Ambeed
Warning

A140778-1G A140778-250MG
77549-714EA 886 USD
77549-714 77592-672
3-Bromo-6-fluoro-2,4-dimethylpyridine ≥97%
3-Bromo-6-fluoro-2,4-dimethylpyridine

Precaución: for Research Use Only

Formula: C₇H₇BrFN
MDL Number: MFCD27930892
CAS Number: 871493-04-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR