3-Phenyl-3-(trifluoromethyl)-3H-diazirine ≥90%

Supplier: Ambeed
Danger

A140272-1G A140272-250MG
77549-710EA 126.69 USD
77549-710 77683-225
3-Phenyl-3-(trifluoromethyl)-3H-diazirine ≥90%
3-Phenyl-3-(trifluoromethyl)-3H-diazirine

Precaución: For research use only.

Formula: C₈H₅F₃N₂
MW: 186.14 g/mol
MDL Number: MFCD22041649
CAS Number: 73899-14-6
UN: 2924
ADR: 3,8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR