Rel-potassium ((1R,2R)-2-(ethoxycarbonyl)cyclopropyl)trifluoroborate 97%

Supplier: Ambeed
Warning

A1402653-1G A1402653-5G A1402653-250MG
77549-708EA 198.79 USD
77549-708 77566-822 77597-144
Rel-potassium ((1R,2R)-2-(ethoxycarbonyl)cyclopropyl)trifluoroborate 97%
Rel-potassium ((1R,2R)-2-(ethoxycarbonyl)cyclopropyl)trifluoroborate

Precaución: For research use only.

Formula: C₆H₉BF₃KO₂
MW: 220.390 g/mol
MDL Number: MFCD31617826
CAS Number: 1612792-88-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR