Potassium(dimethylaminomethyl)trifluoroborate ≥98%

Supplier: Ambeed
Warning

A139887-1G A139887-250MG A139887-5G
77549-700EA 26 USD
77549-700 77891-969 77891-970
Potassium(dimethylaminomethyl)trifluoroborate ≥98%
Potassium (dimethylaminomethyl)trifluoroborate

Precaución: for Research Use Only

Formula: C₃H₈BF₃KN
MDL Number: MFCD10700163
CAS Number: 1150655-04-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR