(3-(tert-Butoxy)-2',6'-diisopropyl-6-methoxy-[1,1'-biphenyl]-2-yl)dicyclohexylphosphane ≥98%

Supplier: Ambeed
Warning

A1376676-1G A1376676-100MG A1376676-250MG A1376676-5G
77549-682EA 803 USD
77549-682 77597-056 77597-058 77867-727
(3-(tert-Butoxy)-2',6'-diisopropyl-6-methoxy-[1,1'-biphenyl]-2-yl)dicyclohexylphosphane ≥98%
(3-(tert-Butoxy)-2',6'-diisopropyl-6-methoxy-[1,1'-biphenyl]-2-yl)dicyclohexylphosphane

Precaución: for Research Use Only

Formula: C₃₅H₅₃O₂P
MW: 536.7679 g/mol
MDL Number: MFCD32857529
CAS Number: 2489243-29-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR