1-Bromo-4-chlorodibenzo[b,d]furan ≥97%

Supplier: Ambeed
Warning

A1376457-5G A1376457-1G
77549-680EA 188.49 USD
77549-680 77621-954
1-Bromo-4-chlorodibenzo[b,d]furan ≥97%
1-Bromo-4-chlorodibenzo[b,d]furan

Precaución: for Research Use Only

Formula: C₁₂H₆BrClO
MW: 281.5324 g/mol
MDL Number: MFCD32669960
CAS Number: 2252237-87-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR